Assay Details
Gene Symbol: NUP35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266251.1 | NP_001253180.1 | 238 | 119 | ||
XM_015110494.2 | XP_014965980.1 | 276 | 119 | |||
GenBank mRNA | JU320089.1 | - | 271 | 119 | ||
JU474324.1 | - | 238 | 119 | |||
JV044824.1 | - | 238 | 119 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
