Assay Details
Gene Symbol: MAPK1IP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260990.1 | NP_001247919.1 | 153 | 159 | ||
GenBank mRNA | JU472902.1 | - | 145 | 159 | ||
JV043762.1 | - | 153 | 159 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001260990.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
