Assay Details
| Gene Symbol: PRDX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260933.1 | NP_001247862.1 | 195 | 138 | ||
| GenBank mRNA | JU475780.1 | - | 143 | 138 | ||
| JV043909.1 | - | 195 | 138 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PRDX1
Genomic Map