Assay Details
Gene Symbol: USP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015120779.2 | XP_014976265.2 | 901 | 151 | ||
XM_015120780.2 | XP_014976266.1 | 1491 | 151 | |||
XM_028837452.1 | XP_028693285.1 | 1569 | 151 | |||
XM_028837453.1 | XP_028693286.1 | 1569 | 151 | |||
XM_028837454.1 | XP_028693287.1 | 1272 | 151 | |||
XM_028837455.1 | XP_028693288.1 | 467 | 151 | |||
GenBank mRNA | JU333379.1 | - | 785 | 151 | ||
JU474676.1 | - | 768 | 151 | |||
JV046441.1 | - | 761 | 151 | |||
JV608796.1 | - | 857 | 151 | |||
JV645134.1 | - | 810 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
