Assay Details
| Gene Symbol: BICDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028829965.1 | XP_028685798.1 | 766 | 74 | ||
| XM_028829966.1 | XP_028685799.1 | 1278 | 74 | |||
| XM_028829967.1 | XP_028685800.1 | 766 | 74 | |||
| XM_028829968.1 | XP_028685801.1 | 766 | 74 | |||
| GenBank mRNA | JV598441.1 | - | 676 | 74 | ||
| JV701326.1 | - | 672 | 74 | |||
More Information
Set Membership: |
|

BICDL1
Genomic Map