Assay Details
Gene Symbol: LOC711508 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015137480.2 | XP_014992966.2 | 659 | 160 | ||
XM_015137481.2 | XP_014992967.2 | 280 | 160 |
Gene Symbol: LOC719437 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015112002.2 | XP_014967488.1 | 357 | 160 | ||
XM_015112017.2 | XP_014967503.1 | 357 | 160 | |||
GenBank mRNA | JU325684.1 | - | 167 | 160 |

More Information
Set Membership: |
![]() ![]() ![]() |
