Assay Details
Gene Symbol: CYCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028845948.1 | XP_028701781.1 | 122 | 143 | ||
GenBank mRNA | JU321674.1 | - | 115 | 143 | ||
JU472428.1 | - | 111 | 143 | |||
JV668536.1 | - | 146 | 143 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028845948.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
