Assay Details
Gene Symbol: ACTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266940.1 | NP_001253869.1 | 877 | 119 | ||
XM_015112506.2 | XP_014967992.1 | 2671 | 119 | |||
XM_028831444.1 | XP_028687277.1 | 927 | 119 | |||
GenBank mRNA | JU332095.1 | - | 902 | 119 | ||
JU332096.1 | - | 887 | 119 | |||
JV045835.1 | - | 882 | 119 | |||
JV045836.1 | - | 882 | 119 | |||
JV045837.1 | - | 882 | 119 | |||
JV635894.1 | - | 877 | 119 | |||
JV667322.1 | - | 892 | 119 | |||
JV704862.1 | - | 943 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
