Assay Details
Gene Symbol: TUBG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261011.1 | NP_001247940.1 | 734 | 126 | ||
GenBank mRNA | JU471734.1 | - | 536 | 126 | ||
JV044189.1 | - | 734 | 126 | |||
JV635408.1 | - | 538 | 126 |
Gene Symbol: LOC100429346 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU320333.1 | - | 720 | 126 | ||
JU471733.1 | - | 536 | 126 | |||
JV046772.1 | - | 734 | 126 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
