Assay Details
Gene Symbol: PPY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193429.3 | NP_001180358.1 | 267 | 129 | ||
XM_015119756.2 | XP_014975242.1 | 397 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
