Assay Details
Gene Symbol: RTRAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266427.1 | NP_001253356.1 | 144 | 102 | ||
GenBank mRNA | JU334946.1 | - | 151 | 102 | ||
JU475950.1 | - | 166 | 102 | |||
JV047339.1 | - | 144 | 102 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
