Assay Details
Gene Symbol: AK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266135.1 | NP_001253064.1 | 598 | 108 | ||
XM_028834219.1 | XP_028690052.1 | 580 | 108 | |||
XM_028834220.1 | XP_028690053.1 | 446 | 108 | |||
GenBank mRNA | JU334741.1 | - | 675 | 108 | ||
JU475786.1 | - | 683 | 108 | |||
JV047215.1 | - | 598 | 108 | |||
JV047216.1 | - | 598 | 108 | |||
JV047217.1 | - | 598 | 108 | |||
JV583313.1 | - | 702 | 108 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
