Assay Details
Gene Symbol: CMTM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266122.1 | NP_001253051.1 | 186 | 136 | ||
GenBank mRNA | JU335150.1 | - | 205 | 136 | ||
JU473492.1 | - | 231 | 136 | |||
JV045281.1 | - | 186 | 136 | |||
JV654731.1 | - | 247 | 136 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
