Assay Details
| Gene Symbol: RBM17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257732.1 | NP_001244661.1 | 1208 | 155 | ||
| XM_028853492.1 | XP_028709325.1 | 1745 | 155 | |||
| GenBank mRNA | JU320162.1 | - | 1208 | 155 | ||
| JU473466.1 | - | 1175 | 155 | |||
More Information
Set Membership: |
|

RBM17
Genomic Map