Assay Details
Gene Symbol: RLIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266839.1 | NP_001253768.1 | 2647 | 139 | ||
XM_015127695.2 | XP_014983181.1 | 2727 | 139 | |||
GenBank mRNA | JU472630.1 | - | 2647 | 139 | ||
JU472631.1 | - | 2647 | 139 | |||
JU472632.1 | - | 2647 | 139 | |||
JU472633.1 | - | 2647 | 139 | |||
JU472634.1 | - | 2647 | 139 | |||
JU472635.1 | - | 2647 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
