Assay Details
| Gene Symbol: RAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133268.2 | XP_014988754.1 | 1227 | 155 | ||
| XM_015133269.2 | XP_014988755.1 | 1170 | 155 | |||
| GenBank mRNA | JU476624.1 | - | 1107 | 155 | ||
| JU476625.1 | - | 1164 | 155 | |||
| JV044974.1 | - | 1116 | 155 | |||
| JV657715.1 | - | 1085 | 155 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RAC1
Genomic Map