Assay Details
| Gene Symbol: ZNF649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124727.2 | XP_014980213.1 | 3029 | 174 | ||
| XM_015124728.2 | XP_014980214.1 | 3322 | 174 | |||
| GenBank mRNA | JU347423.1 | - | 1273 | 174 | ||
| JU491855.1 | - | 1153 | 174 | |||
More Information
Set Membership: |
|

ZNF649
Genomic Map