Assay Details
| Gene Symbol: ZSCAN5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193995.2 | NP_001180924.2 | 1838 | 95 | ||
| XM_015124965.2 | XP_014980451.2 | 3549 | 95 | |||
| XM_015124966.2 | XP_014980452.2 | 2156 | 95 | |||
| XM_028838428.1 | XP_028694261.1 | 2158 | 95 | |||
| GenBank mRNA | JU493306.1 | - | 850 | 95 | ||
More Information
Set Membership: |
|

ZSCAN5A
Genomic Map