Assay Details
Gene Symbol: NIFK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU336720.1 | - | 1078 | 133 | ||
JU477501.1 | - | 1092 | 133 | |||
JV725086.1 | - | 1102 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
