Assay Details
Gene Symbol: BOLA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194470.3 | NP_001181399.2 | 482 | 123 | ||
XM_015125897.2 | XP_014981383.2 | 558 | 123 | |||
XM_028840539.1 | XP_028696372.1 | 116 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() |
