Assay Details
Gene Symbol: GAL3ST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266653.1 | NP_001253582.1 | 1467 | 155 | ||
XM_015150005.2 | XP_015005491.2 | 1602 | 155 | |||
XM_028827392.1 | XP_028683225.1 | 1677 | 155 | |||
XM_028827393.1 | XP_028683226.1 | 3209 | 155 | |||
XM_028827394.1 | XP_028683227.1 | 1632 | 155 | |||
GenBank mRNA | JU337198.1 | - | 1467 | 155 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
