Assay Details
Gene Symbol: SUMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015120251.2 | XP_014975737.1 | 87 | 95 | ||
GenBank mRNA | JU320065.1 | - | 46 | 95 | ||
JU320066.1 | - | 46 | 95 | |||
JU336034.1 | - | 46 | 95 | |||
JU476900.1 | - | 45 | 95 | |||
JV048010.1 | - | 62 | 95 | |||
JV048011.1 | - | 62 | 95 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015120251.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
