Assay Details
Gene Symbol: NXT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194163.2 | NP_001181092.1 | 104 | 144 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194163.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
