Assay Details
Gene Symbol: LOC699924 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015115204.2 | XP_014970690.1 | 2150 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
