Assay Details
Gene Symbol: TMSB15A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001082206.4 | XP_001082206.2 | 615 | 101 | ||
GenBank mRNA | JU470884.1 | - | 581 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
