Assay Details
| Gene Symbol: ZNF552 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028839512.1 | XP_028695345.1 | 1580 | 85 | ||
| XM_028839513.1 | XP_028695346.1 | 2248 | 85 | |||
| GenBank mRNA | JU493629.1 | - | 1627 | 85 | ||
More Information
Set Membership: |
|

ZNF552
Genomic Map