Assay Details
| Gene Symbol: RTP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001103282.3 | XP_001103282.1 | 2702 | 141 | ||
| XM_015132436.2 | XP_014987922.1 | 2269 | 141 | |||
| XM_015132437.2 | XP_014987923.1 | 2142 | 141 | |||
More Information
Set Membership: |
|

RTP1
Genomic Map