Assay Details
Gene Symbol: H2AFZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU319711.1 | - | 52 | 80 | ||
JU471434.1 | - | 45 | 80 | |||
JV044028.1 | - | 56 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
