Assay Details
Gene Symbol: LOC707129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015119648.2 | XP_014975134.1 | 165 | 123 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015119648.2 transcripts.
Set Membership: |
![]() ![]() |
