Assay Details
Gene Symbol: LOC705939 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001105203.3 | XP_001105203.1 | 54 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001105203.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
