Assay Details
| Gene Symbol: ETF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266474.2 | NP_001253403.1 | 2339 | 93 | ||
| XM_015140903.2 | XP_014996389.1 | 2453 | 93 | |||
| GenBank mRNA | JU333360.1 | - | 2312 | 93 | ||
| JU472872.1 | - | 2330 | 93 | |||
| JV044497.1 | - | 2343 | 93 | |||
More Information
Set Membership: |
|

ETF1
Genomic Map