Assay Details
Gene Symbol: CDK2AP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266586.1 | NP_001253515.1 | 268 | 107 | ||
XM_015113520.2 | XP_014969006.1 | 420 | 107 | |||
GenBank mRNA | JU332464.1 | - | 328 | 107 | ||
JU332465.1 | - | 328 | 107 | |||
JU473931.1 | - | 333 | 107 | |||
JU473932.1 | - | 333 | 107 | |||
JV045874.1 | - | 268 | 107 | |||
JV045875.1 | - | 268 | 107 | |||
JV705051.1 | - | 352 | 107 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001266586.1, XM_015113520.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
