Assay Details
| Gene Symbol: LILRA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040675.1 | NP_001035765.1 | 268 | 139 | ||
| GenBank mRNA | DQ155430.1 | - | 268 | 139 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LILRA3
Genomic Map