Assay Details
Gene Symbol: DNTTIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015145331.2 | XP_015000817.1 | 1648 | 138 | ||
XM_015145336.2 | XP_015000822.1 | 2915 | 138 | |||
GenBank mRNA | JU334248.1 | - | 1604 | 138 | ||
JV059384.1 | - | 1604 | 138 | |||
JV579987.1 | - | 1598 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
