Assay Details
| Gene Symbol: CX3CL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032881.1 | NP_001028053.1 | 192 | 75 | ||
| GenBank mRNA | AF449286.1 | - | 192 | 75 | ||
| JV478093.1 | - | 265 | 75 | |||
More Information
Set Membership: |
|

CX3CL1
Genomic Map