Assay Details
| Gene Symbol: IRF2BPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047124.3 | NP_001040589.2 | 2979 | 113 | ||
| GenBank mRNA | JU327194.1 | - | 1854 | 113 | ||
| JV658655.1 | - | 1783 | 113 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

IRF2BPL
Genomic Map