Assay Details
| Gene Symbol: MAMU-B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048245.1 | NP_001041710.1 | 351 | 158 | ||
| GenBank mRNA | AF157402.1 | - | 351 | 158 | ||
| AJ556885.2 | - | 307 | 158 | |||
| AM902546.1 | - | 307 | 158 | |||
| AM902547.1 | - | 307 | 158 | |||
| AM902548.1 | - | 307 | 158 | |||
| EF219481.1 | - | 307 | 158 | |||
| EF372278.1 | - | 340 | 158 | |||
| EU589241.1 | - | 340 | 158 | |||
| EU934763.1 | - | 340 | 158 | |||
| HG813266.1 | - | 340 | 158 | |||
| HM594292.1 | - | 340 | 158 | |||
| LM608033.1 | - | 300 | 158 | |||
| LM608034.1 | - | 300 | 158 | |||
| LN899678.1 | - | 340 | 158 | |||
| LN899679.1 | - | 340 | 158 | |||
More Information
Set Membership: |
|

MAMU-B
Genomic Map