Assay Details
Gene Symbol: LOC106996676 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318353.1 | NP_001305282.1 | 640 | 78 | ||
XM_028842625.1 | XP_028698458.1 | 78 | ||||
GenBank mRNA | AB540194.1 | - | 606 | 78 | ||
AJ556902.1 | - | 78 | ||||
AM902576.1 | - | 607 | 78 | |||
AM902577.1 | - | 607 | 78 | |||
EF580171.1 | - | 78 | ||||
EF581085.1 | - | 631 | 78 | |||
GU592094.1 | - | 631 | 78 | |||
GU592095.1 | - | 631 | 78 | |||
JX442263.1 | - | 640 | 78 | |||
KC205333.1 | - | 78 | ||||
KC205334.1 | - | 78 | ||||
KC205335.1 | - | 78 | ||||
KC205351.1 | - | 478 | 78 | |||
KC205352.1 | - | 478 | 78 | |||
KC205354.1 | - | 478 | 78 | |||
KC205355.1 | - | 478 | 78 | |||
KC205357.1 | - | 478 | 78 | |||
KC205362.1 | - | 78 | ||||
KC205363.1 | - | 478 | 78 | |||
KC205364.1 | - | 478 | 78 | |||
KC205395.1 | - | 640 | 78 | |||
KF791289.1 | - | 640 | 78 | |||
KF855184.1 | - | 78 | ||||
KF855185.1 | - | 78 | ||||
KF855197.1 | - | 640 | 78 | |||
KF855198.1 | - | 640 | 78 | |||
LN899695.1 | - | 78 | ||||
LN899696.1 | - | 78 | ||||
LN899710.1 | - | 640 | 78 | |||
LN899711.1 | - | 640 | 78 | |||
LN899712.1 | - | 640 | 78 | |||
LN899713.1 | - | 640 | 78 | |||
LT623031.1 | - | 640 | 78 | |||
LT623032.1 | - | 640 | 78 | |||
LT623033.1 | - | 640 | 78 | |||
LT623034.1 | - | 640 | 78 | |||
LT623041.1 | - | 640 | 78 | |||
LT908130.1 | - | 78 | ||||
LT908856.1 | - | 640 | 78 | |||
LT908857.1 | - | 640 | 78 |
Gene Symbol: LOC100424348 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842612.1 | XP_028698445.1 | 78 | |||
XM_028842613.1 | XP_028698446.1 | 78 | ||||
XM_028842614.1 | XP_028698447.1 | 78 | ||||
GenBank mRNA | AB477399.1 | - | 78 | |||
AB540197.1 | - | 78 | ||||
AB540204.1 | - | 78 | ||||
AB540210.1 | - | 78 | ||||
AJ556907.1 | - | 78 | ||||
EU363224.1 | - | 78 | ||||
EU934762.1 | - | 78 | ||||
EU934766.1 | - | 631 | 78 | |||
FN396413.1 | - | 78 | ||||
KC205340.1 | - | 78 | ||||
KU850478.1 | - | 78 | ||||
LM608058.1 | - | 78 | ||||
LT623044.1 | - | 78 |
Gene Symbol: LOC106995452 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318357.1 | NP_001305286.1 | 78 | |||
GenBank mRNA | AB540206.1 | - | 78 | |||
EU669871.1 | - | 78 | ||||
KF855182.1 | - | 78 | ||||
KF855183.1 | - | 78 |
Gene Symbol: LOC106996081 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318348.1 | NP_001305277.1 | 78 | |||
GenBank mRNA | AJ556884.1 | - | 78 | |||
AJ556898.1 | - | 78 | ||||
AJ556899.1 | - | 78 | ||||
AM902534.1 | - | 78 | ||||
AM902539.1 | - | 78 | ||||
AM902560.1 | - | 78 | ||||
AM902561.1 | - | 78 | ||||
AM902563.1 | - | 78 | ||||
AM902578.1 | - | 78 | ||||
EF580173.1 | - | 78 | ||||
EF581066.1 | - | 78 | ||||
EU305650.1 | - | 78 | ||||
EU363223.1 | - | 78 | ||||
FN396416.1 | - | 78 | ||||
GU057843.1 | - | 78 | ||||
GU080245.1 | - | 78 | ||||
GU592123.1 | - | 78 | ||||
GU592127.1 | - | 78 | ||||
GU592128.1 | - | 78 | ||||
GU592130.1 | - | 78 | ||||
HG813268.1 | - | 78 | ||||
JX442264.1 | - | 78 | ||||
LN899677.1 | - | 78 | ||||
LN899688.1 | - | 78 | ||||
LN899692.1 | - | 78 | ||||
LN899724.1 | - | 78 | ||||
LT546130.1 | - | 78 | ||||
LT623042.1 | - | 78 | ||||
LT908135.1 | - | 78 | ||||
LT908848.2 | - | 78 |
Gene Symbol: LOC106997893 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028843008.1 | XP_028698841.1 | 78 | |||
XM_028843009.1 | XP_028698842.1 | 78 | ||||
XM_028843010.1 | XP_028698843.1 | 78 | ||||
XM_028843011.1 | XP_028698844.1 | 78 | ||||
GenBank mRNA | AJ556903.1 | - | 78 | |||
AM902579.1 | - | 78 | ||||
EF219478.1 | - | 78 | ||||
EU682516.1 | - | 78 | ||||
FN985469.1 | - | 78 | ||||
GU592156.1 | - | 78 | ||||
JX442275.1 | - | 78 | ||||
KC205336.1 | - | 78 | ||||
KF855202.1 | - | 78 | ||||
LM608070.1 | - | 78 | ||||
LN899697.1 | - | 78 | ||||
LN899716.1 | - | 78 | ||||
LR723074.1 | - | 78 |
Gene Symbol: LOC114676051 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028843007.1 | XP_028698840.1 | 78 |
Gene Symbol: LOC114677644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028847741.1 | XP_028703574.1 | 78 | |||
GenBank mRNA | AJ556908.1 | - | 78 | |||
FN396414.1 | - | 78 | ||||
LT707758.1 | - | 78 |
Gene Symbol: LOC694372 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028847736.1 | XP_028703569.1 | 78 | |||
XM_028847737.1 | XP_028703570.1 | 78 | ||||
XM_028847738.1 | XP_028703571.1 | 78 | ||||
XM_028847739.1 | XP_028703572.1 | 78 | ||||
XM_028847740.1 | XP_028703573.1 | 78 | ||||
GenBank mRNA | AJ556892.1 | - | 78 | |||
AM902551.1 | - | 78 | ||||
EF581081.1 | - | 78 | ||||
EF581082.1 | - | 78 | ||||
FN985459.1 | - | 78 | ||||
FN985460.1 | - | 78 | ||||
GU592149.1 | - | 78 | ||||
GU592150.1 | - | 78 | ||||
KC205326.1 | - | 78 | ||||
LM608036.1 | - | 78 | ||||
LM608037.1 | - | 78 | ||||
LN899682.1 | - | 78 | ||||
LR723055.1 | - | 78 | ||||
LR723056.1 | - | 78 | ||||
LR723062.1 | - | 78 | ||||
LT623002.1 | - | 78 |
Gene Symbol: LOC719250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015129375.2 | XP_014984861.2 | 78 | |||
XM_028842615.1 | XP_028698448.1 | 78 | ||||
XM_028842616.1 | XP_028698449.1 | 78 | ||||
XM_028842617.1 | XP_028698450.1 | 78 | ||||
XM_028842619.1 | XP_028698452.1 | 78 | ||||
XM_028842620.1 | XP_028698453.1 | 78 | ||||
XM_028842621.1 | XP_028698454.1 | 78 | ||||
XM_028842622.1 | XP_028698455.1 | 78 | ||||
XM_028842623.1 | XP_028698456.1 | 78 | ||||
XM_028842624.1 | XP_028698457.1 | 78 |
Gene Symbol: LOC720309 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318351.1 | NP_001305280.1 | 78 | |||
GenBank mRNA | AJ556877.1 | - | 78 | |||
AM902535.1 | - | 78 | ||||
EF580159.1 | - | 78 | ||||
EF580169.1 | - | 78 | ||||
EF581084.1 | - | 78 | ||||
EF611167.1 | - | 78 | ||||
GQ471886.1 | - | 78 | ||||
GU592152.1 | - | 78 | ||||
GU592153.1 | - | 78 | ||||
GU592154.1 | - | 78 | ||||
GU592155.1 | - | 78 | ||||
JX442268.1 | - | 78 | ||||
KF297358.1 | - | 78 | ||||
KF855166.1 | - | 78 | ||||
LT707746.1 | - | 78 |
Gene Symbol: LOC723473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318343.2 | NP_001305272.2 | 78 | |||
XM_015128915.2 | XP_014984401.2 | 78 | ||||
XM_015128917.2 | XP_014984403.2 | 78 | ||||
XM_028842607.1 | XP_028698440.1 | 78 | ||||
XM_028842608.1 | XP_028698441.1 | 78 | ||||
GenBank mRNA | KC205391.1 | - | 78 |
Gene Symbol: MAMU-A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF243179.1 | - | 78 | |||
AJ542574.1 | - | 78 | ||||
AM295888.1 | - | 78 | ||||
EU334729.1 | - | 78 |
Gene Symbol: MAMU-B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048245.1 | NP_001041710.1 | 78 | |||
GenBank mRNA | AF157402.1 | - | 78 | |||
AJ556885.2 | - | 78 | ||||
AM902546.1 | - | 78 | ||||
AM902547.1 | - | 78 | ||||
AM902548.1 | - | 78 | ||||
EF219481.1 | - | 78 | ||||
EF372278.1 | - | 78 | ||||
EU589241.1 | - | 78 | ||||
EU934763.1 | - | 78 | ||||
HG813266.1 | - | 78 | ||||
HM594292.1 | - | 78 | ||||
KC205325.1 | - | 78 | ||||
LM608033.1 | - | 78 | ||||
LM608034.1 | - | 78 | ||||
LN899678.1 | - | 78 | ||||
LN899679.1 | - | 78 |
Gene Symbol: MAMU-B18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114964.1 | NP_001108436.1 | 78 | |||
GenBank mRNA | GU592098.1 | - | 78 | |||
LN899698.1 | - | 78 | ||||
LT546133.1 | - | 78 | ||||
LT623020.1 | - | 78 | ||||
LT908131.1 | - | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
