Assay Details
Gene Symbol: MAMU-DRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044261.3 | NP_001037726.2 | 509 | 86 | ||
XM_028846616.1 | XP_028702449.1 | 556 | 86 | |||
GenBank mRNA | AJ601356.2 | - | 456 | 86 | ||
EU934777.1 | - | 456 | 86 | |||
HM594301.1 | - | 456 | 86 | |||
JQ579552.1 | - | 459 | 86 |
Gene Symbol: LOC100428435 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028847911.1 | XP_028703744.1 | 99 | 86 | ||
GenBank mRNA | JQ579536.1 | - | 459 | 86 | ||
JQ579550.1 | - | 459 | 86 | |||
JQ579554.1 | - | 459 | 86 |
Gene Symbol: LOC705588 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318344.1 | NP_001305273.1 | 456 | 86 | ||
GenBank mRNA | AM690327.1 | - | 456 | 86 | ||
JQ579513.1 | - | 459 | 86 | |||
JQ579515.1 | - | 459 | 86 | |||
JQ579516.1 | - | 459 | 86 |
Gene Symbol: MAMU-DRB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040030.2 | NP_001035119.2 | 503 | 86 | ||
GenBank mRNA | AB185500.1 | - | 456 | 86 | ||
AB185502.1 | - | 456 | 86 | |||
AB185503.1 | - | 456 | 86 | |||
AJ601348.1 | - | 456 | 86 | |||
AJ601350.1 | - | 456 | 86 | |||
AJ601360.2 | - | 456 | 86 | |||
EU934774.1 | - | 456 | 86 | |||
GQ471892.1 | - | 456 | 86 | |||
JV637586.1 | - | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
