Assay Details
Gene Symbol: TAS2R7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080766.1 | NP_001074235.1 | 853 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
