Assay Details
Gene Symbol: TAS2R13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080767.1 | NP_001074236.1 | 852 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
