Assay Details
Gene Symbol: GP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142313.1 | NP_001135785.1 | 209 | 57 | ||
XM_028843221.1 | XP_028699054.1 | 332 | 57 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001142313.1, XM_028843221.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
