Assay Details
Gene Symbol: TTLL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163080.1 | NP_001156552.1 | 1348 | 88 | ||
XM_015130952.2 | XP_014986438.2 | 1931 | 88 | |||
XM_015132143.2 | XP_014987629.2 | 1840 | 88 | |||
XM_015133880.2 | XP_014989366.2 | 1419 | 88 | |||
XM_015134503.2 | XP_014989989.2 | 1482 | 88 | |||
XM_015135345.2 | XP_014990831.2 | 1329 | 88 | |||
XM_028838466.1 | XP_028694299.1 | 1795 | 88 | |||
XM_028838486.1 | XP_028694319.1 | 1851 | 88 | |||
XM_028838564.1 | XP_028694397.1 | 1370 | 88 | |||
GenBank mRNA | FN391584.1 | - | 1348 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
