Assay Details
Gene Symbol: NRARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028834544.1 | XP_028690377.1 | 1450 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
