Assay Details
Gene Symbol: ACOT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171950.1 | NP_001165421.1 | 105 | 67 | ||
GenBank mRNA | CK231547.1 | - | 105 | 67 | ||
JU332310.1 | - | 304 | 67 | |||
JU471589.1 | - | 213 | 67 | |||
JV047637.1 | - | 314 | 67 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
