Assay Details
| Gene Symbol: PEG10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172421.1 | NP_001165892.2 | 2652 | 93 | ||
| NM_001172422.1 | NP_001165893.2 | 2652 | 93 | |||
| GenBank mRNA | JV703365.1 | - | 2906 | 93 | ||
More Information
Set Membership: |
|

PEG10
Genomic Map