Assay Details
| Gene Symbol: SBDS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133370.2 | XP_014988856.1 | 510 | 101 | ||
| GenBank mRNA | JU319987.1 | - | 372 | 101 | ||
| JU473661.1 | - | 390 | 101 | |||
| JV044946.1 | - | 391 | 101 | |||
More Information
Set Membership: |
|

SBDS
Genomic Map