Assay Details
Gene Symbol: OR5A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193891.2 | NP_001180820.1 | 954 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
