Assay Details
| Gene Symbol: NAA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193901.2 | NP_001180830.1 | 881 | 67 | ||
| GenBank mRNA | JV045457.1 | - | 775 | 67 | ||
| JV636419.1 | - | 846 | 67 | |||
More Information
Set Membership: |
|

NAA11
Genomic Map