Assay Details
| Gene Symbol: CCDC122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193957.1 | NP_001180886.1 | 557 | 197 | ||
| XM_015120902.2 | XP_014976388.1 | 1018 | 197 | |||
| XM_028837126.1 | XP_028692959.1 | 2745 | 197 | |||
| XM_028837127.1 | XP_028692960.1 | 1001 | 197 | |||
| XM_028837128.1 | XP_028692961.1 | 856 | 197 | |||
| XM_028837129.1 | XP_028692962.1 | 879 | 197 | |||
| XM_028837130.1 | XP_028692963.1 | 718 | 197 | |||
| XM_028837131.1 | XP_028692964.1 | 596 | 197 | |||
| XM_028837132.1 | XP_028692965.1 | 2445 | 197 | |||
| XM_028837133.1 | XP_028692966.1 | 638 | 197 | |||
More Information
Set Membership: |
|

CCDC122
Genomic Map